Learn More
4-Propionamidophenol (Acetaminophen Impurity B), TRC
Product Code. 30134835
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134835 100 mg 100mg
17911175 250 mg 250mg
17952225 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134835 Supplier Toronto Research Chemicals Supplier No. TRCP783500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1693-37-4
Molecular Formula C9 H11 N O2
Molecular Weight (g/mol) 165.19
Synonym Propanamide, N-(4-hydroxyphenyl)-, Propionanilide, 4'-hydroxy- (7CI,8CI), N-(4-Hydroxyphenyl)propanamide, 4-(Propionylamino)phenol, 4-Propionamidophenol, 4'-Hydroxypropionanilid, 4'-Hydroxypropionanilide, N-(4-Hydroxyphenyl)propionamide, N-(p-Hydroxyphenyl)propionamide, Parapropamol, Parapropanol, Solvodol, p-Hydroxypropionanilide, p-Propionamidophenol, N-Propionyl-4-aminophenol, Paracetamol Imp. B (EP)
IUPAC Name N-(4-hydroxyphenyl)propanamide
SMILES CCC(=O)Nc1ccc(O)cc1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C9H11NO2/c1-2-9(12)10-7-3-5-8(11)6-4-7/h3-6,11H,2H2,1H3,(H,10,12)
Formula Weight 165.079
Percent Purity >95%
Chemical Name or Material 4-Propionamidophenol
Grade HPLC