Learn More
4-Pregnen-17alpha-ol-3,20-dione-2,2,4,6,6,21,21,21-d8, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 17 A-Hydroxyprogesterone-D8 (2,2,4,6,6,21,21,21-D8) |
| CAS | 850023-80-2 |
| Molecular Formula | C21 2H8 H22 O3 |
| Quantity | 0.05 g |
| Synonym | Pregn-4-ene-3,20-dione-2,2,4,6,6,21,21,21-d8, 17-hydroxy-, 17α-Hydroxyprogesterone-D8 (2,2,4,6,6,21,21,21-D8), 2,2,4,6,6,21,21,21-d8-17α-Hydroxyprogesterone |
| InChI Formula | InChI=1 S/C21H30O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h12,16-18,24 H,4-11H2,1-3H3/t16-,17+,18+,19+,20+,21+/m1/s1/i1D3,4D2,6D2,12 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@@H]3 [C@H](CC[C@@]4(C)[C@H]3 CC[C@]4(O)C(=O)C([2 H])([2 H])[2 H])[C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 R)-2,2,4,6,6-pentadeuterio-17-hydroxy-10,13-dimethyl-17-(2,2,2-trideuterioacetyl)-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 338.51 |
| Formula Weight | 338.2697 g/mol |
| Show More |