Learn More
4-Phenylbutyric-2,2,3,3-d4 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Phenylbutanoic-2,2,3,3-D4 Acid |
| CAS | 461391-24-2 |
| Molecular Formula | C10 D4 H8 O2 |
| Quantity | 0.05 g |
| Synonym | Benzenebutanoic-α,α,β,β-d4 acid (9 CI), 4-Phenyl-2,2,3,3-tetradeuteriobutanoic acid, 4-Phenylbutanoic acid-d4, 4-Phenylbutyric acid-d4, ω-Phenylbutanoic acid-d4, Phenylbutyric acid-d4, 4-Phenylbutanoic-2,2,3,3-D4 acid, 4-Phenylbutanoic-2,2,3,3-d4 acid |
| InChI Formula | InChI=1 S/C10H12O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6 H,4,7-8H2,(H,11,12)/i4D2,8D2 |
| SMILES | [2 H]C([2 H])(Cc1ccccc1)C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3-tetradeuterio-4-phenylbutanoic acid |
| Molecular Weight (g/mol) | 168.226 |
| Formula Weight | 168.109 g/mol |
| Show More |