Learn More
4-Phenoxyphenol, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Phenoxyphenol |
| CAS | 831-82-3 |
| Molecular Formula | C12 H10 O2 |
| Quantity | 25 g |
| Synonym | Phenol','p-phenoxy- (6CI,7CI,8CI), 4-Phenoxyphenol, 4-Hydroxydiphenyl ether, Hydroquinone monophenyl ether, NSC 25027, p-Hydroxydiphenyl ether, p-Phenoxyphenol, 4-Hydroxydiphenyl Ether, Hydroquinone Monophenyl Ether, NSC 25027, p-Hydroxydiphenyl Ether, p-Phenoxyphenol, Phenol','p-phenoxy- (6CI,7CI,8CI) |
| InChI Formula | InChI=1S/C12H10O2/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9,13H |
| SMILES | Oc1ccc(Oc2ccccc2)cc1 |
| IUPAC Name | 4-phenoxyphenol |
| Molecular Weight (g/mol) | 186.21 |
| Formula Weight | 186.0681 |