missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)benzyl bromide, 97%

Product Code. 15417207
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417207 1 g 1g
15427207 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417207 Supplier Thermo Scientific Alfa Aesar Supplier No. H34263.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1126969-29-6 | C7H6BrF5S | 297.081 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126969-29-6
Molecular Formula C7H6BrF5S
Molecular Weight (g/mol) 297.081
MDL Number MFCD16652420
InChI Key BFHGXKUYFXAMOZ-UHFFFAOYSA-N
Synonym 4-pentafluorosulfur benzyl bromide, 1-bromomethyl-4-pentafluoro-??-sulfanyl benzene, 4-pentafluorothio benzyl bromide
PubChem CID 58360671
IUPAC Name [4-(bromomethyl)phenyl]-pentafluoro-$l^{6}-sulfane
SMILES C1=CC(=CC=C1CBr)S(F)(F)(F)(F)F

Specifications

Melting Point 42°C to 44°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 297.08
Percent Purity 97%
Chemical Name or Material 4-(Pentafluorothio)benzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.