Learn More
4-O-beta-D-Mannopyranosyl-D-glucopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 4-O-beta-D-Mannopyranosyl-D-glucopyranoside |
| CAS | 29276-55-9 |
| Molecular Formula | C12 H22 O11 |
| Quantity | 25 mg |
| Synonym | D-Glucopyranose, 4-O-β-D-mannopyranosyl-, Glucopyranose, 4-O-β-D-mannopyranosyl- (6CI), Glucopyranose, 4-O-β-D-mannopyranosyl-, D- (8CI), 4-O-β-D-Mannopyranosyl-D-glucopyranose, 4-O-(β-D-Mannopyranosyl)-D-glucopyranose, 4-O-β-D-Mannopyranosyl-D-glucopyranoside |
| InChI Formula | InChI=1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5-,6+,7-,8-,9+,10-,11?,12+/m1/s1 |
| SMILES | OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@@H](O)[C@@H](O)[C@@H]1O |
| IUPAC Name | (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
| Molecular Weight (g/mol) | 342.3 |
| Formula Weight | 342.1162 |