Learn More
Melford 4-Nitrophenyl Phenylphosphonate
Product Code. 30018624
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. 30018624
Brand: Melford N814701.0
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
  • CAS Number: 57072-35-2
  • Molecular Formula: C12H10NO5P
  • Molecular Weight: 279.2 g/mol
Specifications
Quantity 1 g