Learn More
Melford 4-Nitrophenyl-β-D-glucopyranoside
Product Code. p-200087160
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 30018622
Brand: Melford N810901.0
Spectrophotometric substrate for detection of β-D-Glucosidase.
Spectrophotometric substrate for detection of β-D-Glucosidase.
Free PNP: <200ppm
Specifications
Quantity 1 g