Learn More
4-Nitrophenethyl alcohol, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 100-27-6 |
| Molecular Formula | C8H9NO3 |
| MDL Number | MFCD00010202 |
| Quantity | 5 g |
| SMILES | O=[N+](C1=CC=C(C=C1)CCO)[O-] |
| Molecular Weight (g/mol) | 167.16 |
| Percent Purity | 99% |
| Chemical Name or Material | 4-Nitrophenethyl alcohol |