Learn More
4-Nitro Sulfamethoxazole, TRC
Product Code. 30135518
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135518 50 mg 50mg
17964655 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135518 Supplier Toronto Research Chemicals Supplier No. TRCN54900050MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29699-89-6
Molecular Formula C10H9N3O5S
Molecular Weight (g/mol) 283.26
Synonym N-(5-Methyl-3-isoxazolyl)-4-nitrobenzenesulfonamide, N-(5-Methylisoxazol-3-yl)-4-nitrobenzenesulfonamide, Nitrosulfamethoxazole;
IUPAC Name N-(5-methylisoxazol-3-yl)-4-nitrobenzenesulfonamide
SMILES O=S(C1=CC=C([N+]([O-])=O)C=C1)(NC2=NOC(C)=C2)=O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C10H9N3O5S/c1-7-6-10(11-18-7)12-19(16,17)9-4-2-8(3-5-9)13(14)15/h2-6H,1H3,(H,11,12)
Formula Weight 383.03
Percent Purity >95%
Chemical Name or Material 4-Nitro Sulfamethoxazole
Grade HPLC