Learn More
4-Nitro-2-(trifluoromethyl)benzonitrile, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 320-47-8 |
| Molecular Formula | C8H3F3N2O2 |
| MDL Number | MFCD01075729 |
| Quantity | 250 mg |
| SMILES | [O-][N+](=O)C1=CC(=C(C=C1)C#N)C(F)(F)F |
| Molecular Weight (g/mol) | 216.12 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Nitro-2-(trifluoromethyl)benzonitrile |