Learn More
4-n-Propylphenol-d12, CDN
Product Code. 30315541
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315541
Brand: CDN CDND53420.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Propylphenol-D12
CAS 352431-21-1
Molecular Formula C9 D12 O
Quantity 0.25 g
Synonym Phen-2,3,5,6-d4-ol-d, 4-(propyl-d7)- (9 CI), 4-(Propyl-d7)phen-2,3,5,6-d4-ol-d, 4-n-Propylphenol-d12, 4-Propylphenol-D12
InChI Formula InChI=1 S/C9H12O/c1-2-3-8-4-6-9(10)7-5-8/h4-7,10 H,2-3H2,1H3/i1D3,2D2,3D2,4 D,5 D,6 D,7 D/hD
SMILES [2 H]Oc1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC Name 1,2,4,5-tetradeuterio-3-deuteriooxy-6-(1,1,2,2,3,3,3-heptadeuteriopropyl)benzene
Molecular Weight (g/mol) 148.2649
Formula Weight 148.1641 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity