Learn More
4-n-Pentyloxybenzaldehyde, 97%, Thermo Scientific™
Product Code. 11327226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327226 25 g 25g
11317226 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11327226 Supplier Thermo Scientific Alfa Aesar Supplier No. L09089.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5736-91-4
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00014135
InChI Key YAPVGSXODFOBBR-UHFFFAOYSA-N
Synonym 4-pentyloxy benzaldehyde, 4-amyloxybenzaldehyde, p-pentyloxybenzaldehyde, benzaldehyde, 4-pentyloxy, 4-pentyloxybenzaldehyde, benzaldehyde, p-pentyloxy, p-amyloxy benzaldehyde, 4-n-amyloxybenzaldehyde, 4-n-pentyloxybenzaldehyde, p-pentyloxy benzaldehyde
PubChem CID 79815
IUPAC Name 4-pentoxybenzaldehyde
SMILES CCCCCOC1=CC=C(C=C1)C=O
Specifications
Density 1.019
Boiling Point 144°C to 147°C (1 mmHg)
Refractive Index 1.534
Quantity 25 g
Beilstein 642040
Sensitivity Air Sensitive
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material 4-n-Pentyloxybenzaldehyde