missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexylbenzoyl chloride, 98%

Product Code. 11407587
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11407587

Brand: Thermo Scientific Alfa Aesar A15659.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50606-95-6 | C13H17ClO | 224.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50606-95-6
Molecular Formula C13H17ClO
Molecular Weight (g/mol) 224.73
MDL Number MFCD00000701
InChI Key XRAHLPNMIIAEPP-UHFFFAOYSA-N
Synonym 4-n-hexylbenzoyl chloride, benzoyl chloride, 4-hexyl, p-hexylbenzoyl chloride, 4-n-hexylbenzoylchloride, 4-n-hexyl-benzoyl chloride, 4-hexylbenzoyl chloride, para-n-hexyl benzoic acid chloride, 4-hexylbenzene-1-carbonyl chloride
PubChem CID 170863
IUPAC Name 4-hexylbenzoyl chloride
SMILES CCCCCCC1=CC=C(C=C1)C(Cl)=O

Specifications

Density 1.029
Boiling Point 176°C to 177°C (22 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5256
Quantity 25 g
UN Number UN3265
Beilstein 2208938
Sensitivity Moisture sensitive
Formula Weight 224.73
Percent Purity 98%
Chemical Name or Material 4-n-Hexylbenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.