Learn More
4-Methylumbelliferyl 2-Azido-2-deoxy-4,6-O-phenylmethylene-Alpha-D-galactopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methylumbelliferyl 2-Azido-2-deoxy-4,6-O-phenylmethylene-alpha-D-galactopyranoside |
| CAS | 1147438-59-2 |
| Molecular Formula | C23H21N3O7 |
| Quantity | 5 mg |
| Synonym | 7-[[2-Azido-2-deoxy-4,6-O-[phenylmethylene]-3-O-(2,3,4,6-tetra-O-acetyl-beta-D-galactopyranosyl)-alpha-D-galactopyranosyl]oxy]-4-methyl-2H-1-Benzopyran-2-one |
| InChI Formula | InChI=1S/C23H21N3O7/c1-12-9-18(27)31-16-10-14(7-8-15(12)16)30-23-19(25-26-24)20(28)21-17(32-23)11-29-22(33-21)13-5-3-2-4-6-13/h2-10,17,19-23,28H,11H2,1H3/t17-,19-,20-,21+,22+,23+/m1/s1 |
| SMILES | CC1=CC(=O)Oc2cc(O[C@H]3O[C@@H]4CO[C@@H](O[C@@H]4[C@H](O)[C@H]3N=[N+]=[N-])c5ccccc5)ccc12 |
| IUPAC Name | 7-[[(2S,4aR,6R,7R,8R,8aR)-7-azido-8-hydroxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-4-methylchromen-2-one |
| Molecular Weight (g/mol) | 451.43 |
| Formula Weight | 451.138 |