Learn More
4-(Methylthio)benzeneboronic acid, 97%
Product Code. 11405447
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405447 5 g 5g
11495437 1 g 1g
11415447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11405447 Supplier Thermo Scientific Alfa Aesar Supplier No. B23454.06
Request Bulk or Custom Format
CAS: 98546-51-1 | C7H9BO2S | 168.017 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98546-51-1
Molecular Formula C7H9BO2S
Molecular Weight (g/mol) 168.017
MDL Number MFCD00093410
InChI Key IVUHTLFKBDDICS-UHFFFAOYSA-N
Synonym 4-methylthio phenylboronic acid, 4-thioanisoleboronic acid, 4-methylsulfanyl phenylboronic acid, 4-methylthiophenylboronic acid, 4-methylthio phenyl boronic acid, 4-methylsulfanylphenyl boronic acid, 4-methylthio phenylboronicacid, 4-methylthio benzeneboronic acid, thioanisole-4-boronic acid, 4-methylmercapto benzeneboronic acid
PubChem CID 2733959
IUPAC Name (4-methylsulfanylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)SC)(O)O
Specifications
Melting Point ∼208°C to 212°C
Quantity 5 g
Beilstein 3247219
Formula Weight 168.02
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)benzeneboronic acid