Learn More
4-(Methylsulfonyl)acetophenone, TRC
Product Code. 30132712
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132712 5 g 5g
17981315 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30132712 Supplier Toronto Research Chemicals Supplier No. TRCM3290805G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10297-73-1
Molecular Formula C9 H10 O3 S
Molecular Weight (g/mol) 198.24
Synonym Ethanone, 1-[4-(methylsulfonyl)phenyl]-, Acetophenone, 4'-(methylsulfonyl)- (6CI,7CI,8CI), 1-(4-Methylsulfonylphenyl)ethanone, 1-[4-(Methylsulfonyl)phenyl]ethan-1-one, 4-(Methanesulfonyl)acetophenone, 4-(Methylsulfonyl)propiophenone, 4-Methylsulfonylacetophenone, 4'-(Methylsulfonyl)acetophenone, NSC 403928, p-Acetylphenyl methyl sulfone, p-Methylsulfonylacetophenone
IUPAC Name 1-(4-methylsulfonylphenyl)ethanone
SMILES CC(=O)c1ccc(cc1)S(=O)(=O)C
Specifications
Quantity 5 g
InChI Formula InChI=1S/C9H10O3S/c1-7(10)8-3-5-9(6-4-8)13(2,11)12/h3-6H,1-2H3
Formula Weight 198.0351
Chemical Name or Material 4-Methylsulfonylacetophenone