Learn More
4-Methylphenylthio-Beta-D-galactopyranoside, TRC
Product Code. 30473354
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473354 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30473354 Supplier Toronto Research Chemicals Supplier No. TRCM3259155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 28244-98-6
Molecular Formula C13 H18 O5 S
Quantity 5 g
InChI Formula InChI=1S/C13H18O5S/c1-7-2-4-8(5-3-7)19-13-12(17)11(16)10(15)9(6-14)18-13/h2-5,9-17H,6H2,1H3/t9-,10+,11+,12-,13+/m1/s1
SMILES Cc1ccc(S[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)cc1
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methylphenyl)sulfanyloxane-3,4,5-triol
Molecular Weight (g/mol) 286.34
Formula Weight 286.0875
Chemical Name or Material 4-Methylphenylthio-beta-D-galactopyranoside