Learn More
4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol, TRC
Product Code. 30008436
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008436 25 mg 25mg
30007300 10 mg 10mg
30012790 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008436 Supplier Toronto Research Chemicals Supplier No. TRCM32574025MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76014-81-8
Molecular Formula C10 H15 N3 O2
Molecular Weight (g/mol) 209.24
Synonym 3-Pyridinemethanol, α-[3-(methylnitrosoamino)propyl]-, α-[3-(Methylnitrosoamino)propyl]-3-pyridinemethanol, (±)-NNAL, 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol, 4-(Methylnitrosoamino)-1-(3-pyridyl)-1-butanol, 4-(N-Methyl-N-nitrosamino)-1-(3-pyridyl)butan-1-ol, NNAL, Racemic 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol, Racemic NNAL
IUPAC Name N-(4-hydroxy-4-pyridin-3-ylbutyl)-N-methylnitrous amide
SMILES CN(CCCC(O)c1cccnc1)N=O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C10H15N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8,10,14H,3,5,7H2,1H3
Formula Weight 209.1164
Percent Purity >95%
Chemical Name or Material 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol
Grade HPLC