missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol-1,2’,3’,4’,5’,6’-13C6, TRC

Product Code. 30474835
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
Unit Size:
0.25mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474835 1 mg 1mg
30476477 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30474835 Supplier Toronto Research Chemicals Supplier No. TRCM3257411MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189877-28-8
Molecular Formula 13C6 C4 H15 N3 O2
Molecular Weight (g/mol) 215.2
Synonym α-[3-(Methylnitrosoamino)propyl]-3-pyridinemethanol-13C6, (±)-NNAL-13C6, NNAL-13C6, 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol-1,2',3',4',5',6'-13C6, 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol-13C6
IUPAC Name N-(4-hydroxy-4-(2,3,4,5,6-^{13}C_{5})pyridin-3-yl(4^{13}C)butyl)-N-methylnitrous amide
SMILES CN(CCC[13CH](O)[13c]1[13cH][13cH][13cH]n[13cH]1)N=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C10H15N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8,10,14H,3,5,7H2,1H3/i2+1,4+1,6+1,8+1,9+1,10+1
Formula Weight 215.1366
Chemical Name or Material 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol 1,pyridyl-13C6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.