missing translation for 'onlineSavingsMsg'
Learn More

4-Methylmorpholine N-oxide monohydrate, 98+%

Product Code. 11418596
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
Unit Size:
100g
10g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418596 50 g 50g
11408596 10 g 10g
11408730 25 g 25g
11418730 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11418596 Supplier Thermo Scientific Alfa Aesar Supplier No. A15996.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70187-32-5 | C5H13NO3 | 135.163 g/mol

4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylmorpholine N-oxide monohydrate is used as a solvent to prepare cellulose fibers. It is an oxidant and involved in the catalytic OsO4 oxidation of olefins to cis-1,2-diols. It is also involved in ruthenium catalyzed oxidation of alcohols to aldehydes and ketones.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70187-32-5
Molecular Formula C5H13NO3
Molecular Weight (g/mol) 135.163
MDL Number MFCD00149388
InChI Key WAZPLXZGZWWXDQ-UHFFFAOYSA-N
Synonym 4-methylmorpholine n-oxide monohydrate, 4-methylmorpholine 4-oxide hydrate, 4-methylmorpholine n-oxide hydrate, 4-methylmorpholine 4-oxide xhydrate, unii-087wu2ftq5, n-methylmorpholine n-oxide monohydrate, n-methylmorpholine n-oxide hydrate, n-methylmorpholine-n-oxide monohydrate, 4-methyl-4-oxidomorpholin-4-ium hydrate, morpholine, 4-methyl-, 4-oxide, hydrate
PubChem CID 2724197
IUPAC Name 4-methyl-4-oxidomorpholin-4-ium;hydrate
SMILES C[N+]1(CCOCC1)[O-].O

Specifications

Melting Point 73°C to 76°C
Quantity 50 g
Beilstein 5449441
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 135.17 (117.15 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 4-Methylmorpholine N-oxide monohydrate
compliance-icons
Product Identifier
  • N-Methylmorpholine N-oxide monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N-Methylmorpholine N-oxide monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.