missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcarbazole, TRC

Product Code. 30471628
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471628 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471628 Supplier Toronto Research Chemicals Supplier No. TRCM5931351MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylcarbazole
CAS 3770-48-7
Molecular Formula C13H11N
Quantity 1 mg
Synonym 4-Methyl-9H-carbazole
InChI Formula InChI=1S/C13H11N/c1-9-5-4-8-12-13(9)10-6-2-3-7-11(10)14-12/h2-8,14H,1H3
SMILES Cc1cccc2[nH]c3ccccc3c12
IUPAC Name 4-methyl-9H-carbazole
Molecular Weight (g/mol) 181.23
Formula Weight 181.0891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.