Learn More
4-(Methylamino)-2-(methylthio)pyrimidine-5-methanol, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(Methylamino)-2-(methylthio)pyrimidine-5-methanol |
| CAS | 17759-30-7 |
| Molecular Formula | C7H11N3OS |
| Quantity | 100 mg |
| Synonym | 4-(Methylamino)-2-(methylthio)-5-pyrimidinemethanol, (4-(Methylamino)-2-(methylthio )pyrimidin-5-yl)methanol, (4-Methylamino-2-methanethiopyrimidin-5-yl)methanol, (4-Methylamino-2-methylsulfanylpyrimidin-5-yl)methanol |
| InChI Formula | InChI=1S/C7H11N3OS/c1-8-6-5(4-11)3-9-7(10-6)12-2/h3,11H,4H2,1-2H3,(H,8,9,10) |
| SMILES | CNc1nc(SC)ncc1CO |
| Molecular Weight (g/mol) | 185.25 |