Learn More
4-Methyl-2-oxo-2H-chromene-3-carbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-2-oxo-2H-chromene-3-carbonitrile |
| CAS | 24526-69-0 |
| Molecular Formula | C11H7NO2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C11H7NO2/c1-7-8-4-2-3-5-10(8)14-11(13)9(7)6-12/h2-5H,1H3 |
| SMILES | CC1=C(C(OC2=CC=CC=C12)=O)C#N |
| IUPAC Name | 4-methyl-2-oxochromene-3-carbonitrile |
| Molecular Weight (g/mol) | 185.18 |
| Formula Weight | 185.0477 |