Learn More
4-Methoxybenzyl mercaptan, 98%
Product Code. p-7026515
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445107 5 g 5g
11455107 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11445107 Supplier Thermo Scientific Alfa Aesar Supplier No. B22542.06
Request Bulk or Custom Format
CAS: 6258-60-2 | C8H10OS | 154.23 g/mol
4-Methoxybenzyl mercaptan is used to synthesize many ω-mercapto amino acids with side-chain lengths ranging from 3-5 methylene units. It is used as an internal standard in the Quantitative determination of wine polyfunctional mercaptans at the level of nanogram per liter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzyl mercaptan is used to synthesize many ω-mercapto amino acids with side-chain lengths ranging from 3-5 methylene units. It is used as an internal standard in the Quantitative determination of wine polyfunctional mercaptans at the level of nanogram per liter.
Solubility
Not miscible or difficult to mix in water.
Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agents, acids, bases, air.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 6258-60-2
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00004871
InChI Key PTDVPWWJRCOIIO-UHFFFAOYSA-N
Synonym 4-methoxybenzyl mercaptan, 4-methoxy-alpha-toluenethiol, 4-methoxybenzylmercaptan, 4-methoxyphenyl methanethiol, 4-methoxy benzylmercaptan, 4-methoxybenzenemethanethiol, benzenemethanethiol, 4-methoxy, p-methoxybenzyl mercaptan, p-methoxybenzylthiol, p-methoxybenzylmercaptan
PubChem CID 80407
IUPAC Name (4-methoxyphenyl)methanethiol
SMILES COC1=CC=C(CS)C=C1
Spécification
Density 1.112
Boiling Point 90°C to 95°C (0.5 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.5755
Quantity 5 g
Beilstein 471686
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 154.23
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzyl mercaptan
Afficher plus Afficher moins
RUO – Research Use Only
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.