Learn More
4-Methoxy-2-(trifluoromethyl)benzaldehyde, 98%, BLD Pharm™
Especificaciones
Especificaciones
| CAS | 106312-36-1 |
| Molecular Formula | C9H7F3O2 |
| MDL Number | MFCD01091011 |
| Quantity | 5 g |
| SMILES | O=CC1=CC=C(OC)C=C1C(F)(F)F |
| Molecular Weight (g/mol) | 204.15 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Methoxy-2-(trifluoromethyl)benzaldehyde |