missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2,3,6-trimethylbenzenesulfonyl chloride, tech. 90%

Product Code. 11352647
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352647 5 g 5g
11342647 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352647 Supplier Thermo Scientific Alfa Aesar Supplier No. L11829.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 80745-07-9 | C10H13ClO3S | 248.721 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80745-07-9
Molecular Formula C10H13ClO3S
Molecular Weight (g/mol) 248.721
MDL Number MFCD00038846
InChI Key DWLYVEYCCPEHLX-UHFFFAOYSA-N
Synonym 4-methoxy-2,3,6-trimethylbenzene-1-sulfonyl chloride, 4-methoxy-2,3,6-trimethyl-benzenesulfonyl chloride, mtr chloride, 4-methoxy-2,3,6-trimethylbenzenesulfonylchloride, 4-methoxy-2,3,6-trimethylphenylsulfonylchloride, chloro 4-methoxy-2,3,6-trimethylphenyl sulfone, 4-methoxy-2,3,6-trimethylphenyl sulfonylchloride, 4-methoxy-2,3,6-trimethylbenzensulfonyl chloride, 4-methoxy-2,3,6-trimethylphenylaulfomyl chloride, 4-methoxy-2,3,6-trimethylphenylsulfonyl chloride
PubChem CID 562787
IUPAC Name 4-methoxy-2,3,6-trimethylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C(=C1S(=O)(=O)Cl)C)C)OC

Specifications

Melting Point 56°C to 59°C
Quantity 5 g
UN Number UN3261
Beilstein 4867665
Sensitivity Moisture sensitive
Formula Weight 248.73
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Methoxy-2,3,6-trimethylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.