missing translation for 'onlineSavingsMsg'
Learn More

TdB Labs AB 4 kDa Q-dextran for Ion-exchange Chromatography and Biomolecular Studies

Product Code. p-200113302
Encompass_Preferred
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 30216318

Brand: TdB Labs AB QD4_10g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Q-dextran is a positively charged dextran modified with quaternary ammonium groups. It is used in ion exchange chromatography, biochemical assays, and for studying electrostatic interactions with negatively charged biomolecules.

  • Storage and Stability:
    • Store dry at room temperature, protected from moisture and light.
    • Stable for extended periods under recommended storage conditions without loss of charge properties.
  • Specifications

    Quantity 10 g
    Product Type Modified Dextran
    Product Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.