Learn More
4-Isopropoxyaniline, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 7664-66-6 |
| Molecular Formula | C9H13NO |
| MDL Number | MFCD00059188 |
| Quantity | 10 g |
| SMILES | NC1=CC=C(OC(C)C)C=C1 |
| Molecular Weight (g/mol) | 151.21 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Isopropoxyaniline |