missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™

Product Code. p-3485763
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10235334 250 mg 250mg
10502865 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10235334 Supplier Thermo Scientific Maybridge Supplier No. CC45806CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 887922-92-1
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD08690262
InChI Key TVGVHXVLMROUII-UHFFFAOYSA-N
Synonym 4-isocyanato-1-methyl-1h-indole, 1h-indole, 4-isocyanato-1-methyl, 1-methylindol-4-isocyanate, 1h-indole,4-isocyanato-1-methyl, 1-methyl-1h-indole-4-yl isocyanate
PubChem CID 18525783
IUPAC Name 4-isocyanato-1-methylindole
SMILES CN1C=CC2=C(C=CC=C21)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 4-Isocyanato-1-methyl-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.