Learn More
4'-Isobutylacetophenone, 97%
Product Code. p-7080887
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. 11433334
Brand: Thermo Scientific Alfa Aesar A10624.18
Request Bulk or Custom Format
CAS: 38861-78-8 | C12H16O | 176.259 g/mol
4'-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.
Solubility
Miscible with chloroform and methanol. Slightly miscible with water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition. Incompatible with reducing agents, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 38861-78-8
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00027393
InChI Key KEAGRYYGYWZVPC-UHFFFAOYSA-N
Synonym 4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20
PubChem CID 93214
IUPAC Name 1-[4-(2-methylpropyl)phenyl]ethanone
SMILES CC(C)CC1=CC=C(C=C1)C(=O)C
Specifications
Density 0.952
Flash Point 54°C (129°F)
Refractive Index 1.518
Quantity 50 g
UN Number UN1224
Beilstein 1935275
Solubility Information Miscible with chloroform and methanol. Slightly miscible with water.
Formula Weight 176.27
Percent Purity 97%
Chemical Name or Material 4'-Isobutylacetophenone
compliance-icons
Product Identifier
  • 4'-Isobutylacetophenone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P233-Keep container tightly closed.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .