Learn More
4-Indolylboronic acid, 95%, Thermo Scientific™
Product Code. 15264358 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15264358
Brand: Thermo Scientific Acros 454500050
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Chemical Identifiers
CAS 220465-43-0
Molecular Formula C8H8BNO2
Molecular Weight (g/mol) 160.97
InChI Key USJUQEVUEBCLLR-UHFFFAOYSA-N
Synonym indole-4-boronic acid, 4-boronoindole, 1h-indol-4-yl boronic acid, 1h-indole-4-boronic acid, 4-indoleboronic acid, 4-indolylboronic acid, 4-borono-1h-indole, 1h-indol-4-yl-4-boronic acid, boronic acid, 1h-indol-4-yl, b-1h-indol-4-yl-boronic acid
PubChem CID 2763173
IUPAC Name 1H-indol-4-ylboronic acid
SMILES B(C1=C2C=CNC2=CC=C1)(O)O
Specifications
Melting Point 184.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 160.97
Percent Purity 95%
Chemical Name or Material 4-Indolylboronic acid
compliance-icons
Product Identifier
  • 4-Indolylboronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.