missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxypiperidine-2,2,3,3,4,5,5,6,6-d9, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxypiperidine-d9 |
| CAS | 1219799-39-9 |
| Molecular Formula | C5H2D9NO |
| Quantity | 0.1 g |
| Synonym | 4-Hydroxypiperidine-2,2,3,3,4,5,5,6,6-d9, 4-Piperidin-2,2,3,3,4,5,5,6,6-d9-ol (ACI) |
| InChI Formula | InChI=1 S/C5H11NO/c7-5-1-3-6-4-2-5/h5-7 H,1-4H2/i1D2,2D2,3 D,4D2,5 D/hD |
| SMILES | OC1([2 H])C([2 H])([2 H])C([2 H])([2 H])N([2 H])C([2 H])C1([2 H])[2 H] |
| IUPAC Name | piperidin-1,2,2,3,3,4,5,5,6-d9-4-ol |
| Molecular Weight (g/mol) | 110.2 |
| Formula Weight | 110.14 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction