missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Hydroxyphenyl-3,5-d2)acetic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxyphenylacetic Acid-D4 (acetic-D2,4-hydroxyphenyl-3,5-D2) |
| Molecular Formula | C8 D4 H4 O3 |
| Quantity | 0.1 g |
| Synonym | 4-Hydroxybenzene-3,5-d2-acetic-α,α-d2 acid, 4-Hydroxyphenylacetic acid-D4 (acetic-D2,4-hydroxyphenyl-3,5-D2), 2-(4'-Hydroxyphenyl-3',5'-d2)acetic-2,2-d2 acid, 4-(Carboxymethyl-d2)phenol-2,6-d2, 4'-Hydroxy-3',5'-dideuteriophenylacetic-2,2-d2 acid, (4-Hydroxyphenyl-3,5-d2)acetic-2,2-d2 Acid |
| InChI Formula | InChI=1 S/C8H8O3/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4,9 H,5H2,(H,10,11)/i3D,4 D,5D2 |
| SMILES | [2 H]c1cc(cc([2 H])c1O)C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-(3,5-dideuterio-4-hydroxyphenyl)acetic acid |
| Molecular Weight (g/mol) | 156.172 |
| Formula Weight | 156.0725 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction