Learn More
4'-Hydroxynonanophenone, 96%
Product Code. 11357655
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357655 25 g 25g
11347655 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11357655 Supplier Thermo Scientific Alfa Aesar Supplier No. L02293.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
CAS: 14392-69-9 | C15H22O2 | 234.34 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14392-69-9
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.34
MDL Number MFCD00020174
InChI Key JZBXYOOARNRUME-UHFFFAOYSA-N
Synonym 1-4-hydroxyphenyl-1-nonanone, 1-4-hydroxyphenyl nonan-1-one, p-hydroxynonanophenone, 4'-hydroxynonanophenone, pelargonyl-4-phenol, 1-nonanone,1-4-hydroxyphenyl, 1-nonanone, 1-4-hydroxyphenyl
PubChem CID 167093
IUPAC Name 1-(4-hydroxyphenyl)nonan-1-one
SMILES CCCCCCCCC(=O)C1=CC=C(O)C=C1
Specifications
Melting Point 53°C to 57°C
Quantity 25 g
Beilstein 2103401
Formula Weight 234.34
Percent Purity 96%
Chemical Name or Material 4'-Hydroxynonanophenone