missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzoic-1,2,3,4,5,6-13C6 Acid, TRC

Product Code. 30010959
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010959 10 mg 10mg
30135277 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010959 Supplier Toronto Research Chemicals Supplier No. TRCH80845310MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287399-29-5
Molecular Formula 13C6 C H6 O3
Molecular Weight (g/mol) 144.08
Synonym Benzoic-1,2,3,4,5,6-13C6 acid, 4-hydroxy- (9CI), p-Salicylic acid-13C6 (phenyl-13C6), 4-Carboxyphenol-13C6, 4-Hydroxybenzene-13C6-carboxylic acid, p-Carboxyphenol-13C6, p-Hydroxybenzoic acid-13C6, 4-Hydroxy-1,2,3,4,5,6-13C6-benzoic acid
IUPAC Name 4-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-carboxylic acid
SMILES O[13c]1[13cH][13cH][13c]([13cH][13cH]1)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1
Formula Weight 144.0518
Percent Purity >95%
Chemical Name or Material 4-Hydroxybenzoic acid phenyl-13C6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.