Learn More
4-Hydroxybenzenesulfonic Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxybenzenesulfonic Acid Sodium Salt |
| CAS | 825-90-1 |
| Molecular Formula | C6H5NaO4S |
| Quantity | 50 g |
| Synonym | Sodium 4-Hydroxybenzenesulfonate, Sodium p-Hydroxybenzenesulfonate, Sodium p-Hydroxyphenylsulfonate, Sodium p-Phenolsulfonate, p-Hydroxybenzenesulfonic Acid Monosodium Salt, p-Hydroxybenzenesulfonic Acid Sodium Salt, p-Phenolsulfonic Acid Sodium Salt |
| InChI Formula | InChI=1S/C6H6O4S.Na/c7-5-1-3-6(4-2-5)11(8,9)10;/h1-4,7H,(H,8,9,10);/q;+1/p-1 |
| SMILES | [Na+].Oc1ccc(cc1)S(=O)(=O)[O-] |
| IUPAC Name | sodium;4-hydroxybenzenesulfonate |
| Molecular Weight (g/mol) | 196.16 |
| Formula Weight | 195.9806 |