missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Hydroxy-3'-methoxy-d3-acetophenone, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Acetovanillone-d3 |
| CAS | 80404-23-5 |
| Molecular Formula | C9 D3 H7 O3 |
| Quantity | 0.01 g |
| Synonym | Ethanone, 1-[4-hydroxy-3-(methoxy-d3)phenyl]- (9 CI, ACI), 1-[4-Hydroxy-3-(methoxy-d3)phenyl]ethanone (ACI), 4'-Hydroxy-3'-methoxy-d3-acetophenone, Acetovanillone-d3, Apocynin-d3, 1-(4-Hydroxy-3-methoxyphenyl)ethanone-D3 (methoxy-D3) |
| InChI Formula | InChI=1 S/C9H10O3/c1-6(10)7-3-4-8(11)9(5-7)12-2/h3-5,11 H,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1cc(ccc1O)C(=O)C |
| IUPAC Name | 1-[4-hydroxy-3-(trideuteriomethoxy)phenyl]ethanone |
| Molecular Weight (g/mol) | 169.192 |
| Formula Weight | 169.082 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction