Learn More
4-Hydroxy-2,2,6,6-tetramethylpiperidine-d17-1-15N-1-oxyl, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl-D17-15 N |
| CAS | 90429-66-6 |
| Molecular Formula | C9 D17 H 15 N O2 |
| Quantity | 0.01 g |
| Synonym | 1-Piperidinyl-3,3,4,5,5-d5-1-15 N-oxy, 4-hydroxy-2,2,6,6-tetra(methyl-d3)- (9 CI, ACI), 4-Hydroxy-2,2,6,6-tetra(methyl-d3)-1-piperidinyl-3,3,4,5,5-d5-1-15 N-oxy (ACI), Tanold-15, 4-Hydroxy-2,2,6,6-tetramethylpiperidine-d17-1-15 N-1-oxyl, TEMPOL-d17-15 N, 4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl-D17-15 N, 4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl-D17-15 N, 4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl-D17-15 N |
| InChI Formula | InChI=1 S/C9H18NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h7,11 H,5-6H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7 D,10+1 |
| SMILES | [2 H]C([2 H])([2 H])C1([15 N]([O])C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])(O)C1([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 3,3,4,5,5-pentadeuterio-1-λ1-oxidanyl-2,2,6,6-tetrakis(trideuteriomethyl)(115 N)azinan-4-ol |
| Molecular Weight (g/mol) | 190.3429 |
| Formula Weight | 190.2375 g/mol |
| Show More |