Learn More
4-Formylbenzoic Acid, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 4-Formylbenzoic Acid |
| CAS | 619-66-9 |
| Molecular Formula | C8H6O3 |
| Quantity | 100 g |
| Synonym | Terephthalaldehydic Acid, 4-Carboxybenzaldehyde, 4-Carboxylbenzaldehyde, NSC 15797, Terephthalaldehyde Acid, Terephthaldehydic Acid, p-Benzoic Acid Aldehyde, p-Carboxybenzaldehyde, p-Formylbenzoic Acid |
| InChI Formula | InChI=1S/C8H6O3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-5H,(H,10,11) |
| SMILES | OC(=O)c1ccc(C=O)cc1 |
| Molecular Weight (g/mol) | 150.13 |
| Formula Weight | 150.0317 |