missing translation for 'onlineSavingsMsg'
Learn More

4-fluorothioanisole 97%, Thermo Scientific™

Product Code. 10006282 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10006282 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10006282 Supplier Thermo Scientific Acros Supplier No. 383490010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 371-15-3
CAS Min % 96.0
CAS Max % 100.0
Density 1.1670g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H7FS
Linear Formula FC6H4SCH3
Refractive Index 1.5500 to 1.5520 (20°C, 589nm)
MDL Number MFCD00040829
Quantity 1g
Synonym 4-fluorothioanisole, 1-fluoro-4-methylthio benzene, 4-fluorophenyl methyl sulfide, 4-fluoro thioanisole, 1-fluoro-4-methylsulfanyl-benzene, 1-fluoro-4-methylsulfanyl benzene, p-fluoro methylthio benzene, benzene, 1-fluoro-4-methylthio, 4-fluoro-1-methylthiobenzene, pubchem10641
InChI Key XFUMHENRNCUHOH-UHFFFAOYSA-N
SMILES CSC1=CC=C(C=C1)F
IUPAC Name 1-fluoro-4-methylsulfanylbenzene
Molecular Weight (g/mol) 142.191
PubChem CID 3083565
Formula Weight 142.2
Percent Purity ≥96.0%
Chemical Name or Material 4-Fluorothioanisole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.