missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorophenol-d5, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-Fluorophenol D5 |
| CAS | 1219804-93-9 |
| Molecular Formula | C6 2H5 F O |
| Quantity | 0.5 g |
| Synonym | 4-Fluorophenol D5, 4-Fluorophenol-d5, Perdeuterio-4-fluorophenol, 4-Fluorophenol-1,2,3,5,6-D5 |
| InChI Formula | InChI=1 S/C6H5FO/c7-5-1-3-6(8)4-2-5/h1-4,8 H/i1D,2 D,3 D,4 D/hD |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c(F)c([2 H])c1[2 H] |
| IUPAC Name | 1,2,4,5-tetradeuterio-3-deuteriooxy-6-fluorobenzene |
| Molecular Weight (g/mol) | 117.132 |
| Formula Weight | 117.064 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction