missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzylamine, TRC

Product Code. 30402080
Click to view available options
Quantity:
1 g
100 g
Unit Size:
100g
1g
This item is not returnable. View return policy

Product Code. 30402080

Brand: Toronto Research Chemicals TRCF588510100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluorobenzylamine
CAS 140-75-0
Molecular Formula C7 H8 F N
Quantity 100 g
Synonym Benzenemethanamine, 4-fluoro- (9CI, ACI), 4-Fluorobenzenemethanamine (ACI), Benzylamine, p-fluoro- (7CI, 8CI), (4-Fluorophenyl)methanamine, 1-(4-Fluorophenyl)methanamine, 4-Fluorobenzylamine, [(4-Fluorophenyl)methyl]amine, NSC 158269, p-Fluorobenzylamine, Benzenemethanamine, 4-fluoro- (9CI, ACI), 4-Fluorobenzenemethanamine (ACI), Benzylamine, p-fluoro- (7CI, 8CI), (4-Fluorophenyl)methanamine, 1-(4-Fluorophenyl)methanamine, 4-Fluorobenzylamine, [(4-Fluorophenyl)methyl]amine, NSC 158269, p-Fluorobenzylamine
InChI Formula InChI=1S/C7H8FN/c8-7-3-1-6(5-9)2-4-7/h1-4H,5,9H2
SMILES NCc1ccc(F)cc1
IUPAC Name (4-fluorophenyl)methanamine
Molecular Weight (g/mol) 125.14
Formula Weight 125.0641
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.