missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylbenzoyl Chloride, TRC

Product Code. 30405243
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405243 500 mg -
30404765 50 mg -
30398455 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405243 Supplier Toronto Research Chemicals Supplier No. TRCF600498500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-84-5
Molecular Formula C8H6OFCl
Molecular Weight (g/mol) 172.58
Synonym 3-Methyl-4-fluorobenzoyl Chloride
SMILES Cc1cc(ccc1F)C(=O)Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H6ClFO/c1-5-4-6(8(9)11)2-3-7(5)10/h2-4H,1H3
Formula Weight 172.0091
Chemical Name or Material 4-Fluoro-3-methylbenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.