Learn More
4-Fluoro-3-methylbenzoyl chloride, 97%, Thermo Scientific™
Product Code. 11430787
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430787 5 g 5g
11420787 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11430787 Supplier Thermo Scientific Alfa Aesar Supplier No. B23188.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 455-84-5
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.583
MDL Number MFCD01631426
InChI Key FMFLKTDRPXYUGP-UHFFFAOYSA-N
Synonym 4-fluoro-3-methylbenzoylchloride, 4-fluoro-m-toluoyl chloride, 4-fluoro-3-methyl-benzoyl chloride, benzoyl chloride, 4-fluoro-3-methyl, acmc-1arwr, 5-chlorocarbonyl-2-fluorotoluene, 4-fluoranyl-3-methyl-benzoyl chloride, 4-fluoro-3-methyl benzoyl chloride, 4-fluoro-3-methyl-benzoic acid chloride, 4-fluoro-3-methylbenzoyl chloride
PubChem CID 2774589
IUPAC Name 4-fluoro-3-methylbenzoyl chloride
SMILES CC1=C(C=CC(=C1)C(=O)Cl)F
Specifications
Density 1.22
Boiling Point 110°C to 115°C (20 mmHg)
Flash Point 98°C (208°F)
Odor Pungent
Refractive Index 1.5313
Quantity 5 g
UN Number UN3265
Beilstein 1936532
Sensitivity Moisture sensitive
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylbenzoyl chloride