missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-2-methoxyphenylacetic acid, 97%

Product Code. 11421400
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11421400

Brand: Alfa Aesar™ H26430.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886498-61-9
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.166
MDL Number MFCD06660193
InChI Key TUHNRCPWPMJXQN-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxyphenylacetic acid, 2-4-fluoro-2-methoxyphenyl acetic acid, 4-fluoro-2-methoxy phenylacetic acid, 4-fluoro-2-methoxybenzeneacetic acid, 4-fluoro-2-methoxyphenyl acetic acid
PubChem CID 17750773
IUPAC Name 2-(4-fluoro-2-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)F)CC(=O)O

Specifications

Melting Point 106°C to 110°C
Quantity 1 g
Formula Weight 184.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxyphenylacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.