Learn More
4-Ethynylpyridine Hydrochloride (>85%), TRC
Specifications
Specifications
| Chemical Name or Material | 4-Ethynylpyridine Hydrochloride |
| CAS | 352530-29-1 |
| Molecular Formula | C7 H5 N . H Cl |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C7H5N.ClH/c1-2-7-3-5-8-6-4-7;/h1,3-6H;1H |
| SMILES | Cl.C#Cc1ccncc1 |
| IUPAC Name | 4-ethynylpyridine;hydrochloride |
| Molecular Weight (g/mol) | 139.582 |
| Formula Weight | 139.019 |