missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Ethynylphenylacetonitrile, 95%, Thermo Scientific™

Product Code. 15427628
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15427628

Brand: Alfa Aesar™ H61122.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351002-90-9
Molecular Formula C10H7N
Molecular Weight (g/mol) 141.17
MDL Number MFCD05664204
InChI Key CZUBOLAETAAHLA-UHFFFAOYSA-N
Synonym 4-ethynylphenylacetonitrile, 2-4-ethynylphenyl acetonitrile, 4-ethynylphenyl acetonitrile, benzeneacetonitrile, 4-ethynyl, 4-ethynylbenzeneacetonitrile, ksc497s1l
PubChem CID 16069314
IUPAC Name 2-(4-ethynylphenyl)acetonitrile
SMILES C#CC1=CC=C(CC#N)C=C1

Specifications

Density 1.037
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 5 g
UN Number UN3276
Solubility Information Slightly soluble in water.
Formula Weight 141.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Ethynylphenylacetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.