Learn More
4-Ethylbenzoic acid, 97%
Product Code. 10040540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10040540 25 g Glass bottle Each
10563741 5 g Glass bottle 5g
10042420 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10040540 Supplier Lenz Laborglasintrumente Supplier No. B0496601
CAS: 619-64-7 | C9H10O2 | 150.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS 619-64-7
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002570
InChI Key ZQVKTHRQIXSMGY-UHFFFAOYSA-N
Synonym benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid;
PubChem CID 12086
IUPAC Name 4-ethylbenzoic acid
SMILES CCC1=CC=C(C=C1)C(=O)O
Specifications
CAS Min % 96.0
CAS Max % 100.0
Melting Point 110.0°C to 114.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C2H5C6H4CO2H
Quantity 25 g
Beilstein 09, 529
Solubility Information Solubility in water: slightly soluble
Formula Weight 150.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Ethylbenzoic acid, 97%