Learn More
4-Ethoxyphenylacetylene, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 79887-14-2 |
| Molecular Formula | C10H10O |
| MDL Number | MFCD00173880 |
| Quantity | 100 g |
| SMILES | C(C)OC1=CC=C(C=C1)C#C |
| Molecular Weight (g/mol) | 146.19 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Ethoxyphenylacetylene |